ContactUsLinkComponent
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW of 1 * 50 µG
Catalog # ACRBDP4-H5211-50UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW of 1 * 50 µG
Catalog # ACRBDP4-H5211-50UG
Supplier:  ACROBIOSYSTEMS