ContactUsLinkComponent
Recombinat Protein,Human DR6 TNFRSF21 Protein, Fc Tag,Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It c ontains AA Gln 42 - Leu 350 (Accession AAH17730).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBTN1-H5252-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human DR6 TNFRSF21 Protein, Fc Tag,Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It c ontains AA Gln 42 - Leu 350 (Accession AAH17730).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBTN1-H5252-1MG
Supplier:  ACROBIOSYSTEMS