Auftragserfassung
Germany
ContactUsLinkComponent
Biotinylated Human CD32a (H167), Avitag,His Tag (SPR  BLI verified) (CDA-H82E6) is expressed from human 293 cells (HEK293). It conta ins AA Ala 36 - Ile 218 (Accession P12318-1 (H167)). 1 * 25 µG
Catalog # ACRBCDA-H82E6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD32a (H167), Avitag,His Tag (SPR  BLI verified) (CDA-H82E6) is expressed from human 293 cells (HEK293). It conta ins AA Ala 36 - Ile 218 (Accession P12318-1 (H167)). 1 * 25 µG
Catalog # ACRBCDA-H82E6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together