ContactUsLinkComponent
Biotinylated Human CD16a (V176), Avitag,His Tag (SPR  BLI verified) (CDA-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 200 µG
Catalog # ACRBCDA-H82E9200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD16a (V176), Avitag,His Tag (SPR  BLI verified) (CDA-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 200 µG
Catalog # ACRBCDA-H82E9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together