ContactUsLinkComponent
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag™,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is expresse d from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhist 1 * 25 µG
Catalog # ACRBLA1-H82E3-25UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human LAIR1 CD305 Protein, His,Avitag™,Biotinylated Human LAIR-1, His,Avitag (LA1-H82E3) is expresse d from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession Q6GTX8-1).This protein carries a polyhist 1 * 25 µG
Catalog # ACRBLA1-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together