ContactUsLinkComponent
Biotinylated Mouse VEGF120, His,Avitag (VE0-M82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Acce ssion Q00731-3). 1 * 200 µG
Catalog # ACRBVE0-M82Q2200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse VEGF120, His,Avitag (VE0-M82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Acce ssion Q00731-3). 1 * 200 µG
Catalog # ACRBVE0-M82Q2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together