Cynomolgus CD3ECD3D Heterodimer Protein, His TagFlag Tag, Gln 22 - Asp 117 (CD3E,Q95LI5) and Phe 22 - Ala 105 (CD3D,Q95LI8) was prod uced in human 293 cells (HEK293) at ACROBiosystems. 1 * 1 mg
Catalog # ACRBCDD-C52W4-1MG
Supplier: ACRO Biosystems
Frequently Bought Together