ContactUsLinkComponent
Cynomolgus Rhesus macaque CTLA-4, Fc Tag (CT4-C5256) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Ac cession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homolo 1 * 200 µG
Catalog # ACRBCT4-C5256200UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque CTLA-4, Fc Tag (CT4-C5256) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Ac cession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homolo 1 * 200 µG
Catalog # ACRBCT4-C5256200UG
Supplier:  ACRO Biosystems
Frequently Bought Together