ContactUsLinkComponent
Recombinat Protein,Cynomolgus LILRB4 CD85k ILT3 Protein, Fc Tag,Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 ce lls (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP015297198.1).This protein carries a human IgG1 Fc tag at t 1 * 1 mg
Catalog # ACRBCDK-C5258-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus LILRB4 CD85k ILT3 Protein, Fc Tag,Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 ce lls (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP015297198.1).This protein carries a human IgG1 Fc tag at t 1 * 1 mg
Catalog # ACRBCDK-C5258-1MG
Supplier:  ACROBIOSYSTEMS