ContactUsLinkComponent
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed fr om human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidin 1 * 1 mg
Catalog # ACRBAN3-H5226-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed fr om human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidin 1 * 1 mg
Catalog # ACRBAN3-H5226-1MG
Supplier:  ACROBIOSYSTEMS