Recombinat Protein,Human Annexin A5 Protein, His Tag,Human Annexin A5, His Tag (AN5-H5123) is expressed from E.coli cells. It contai ns AA Met 1 - Asp 320 (Accession P08758-1).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
Catalog # ACRBAN5-H5123-1MG
Lieferant: ACRO Biosystems
Frequently Bought Together