ContactUsLinkComponent
Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 3 43 (Accession NP006496.4). 1 * 100 µG
Catalog # ACRBCD5-H5254100UG
Supplier:  ACRO Biosystems
Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 3 43 (Accession NP006496.4). 1 * 100 µG
Catalog # ACRBCD5-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together