ContactUsLinkComponent
Recombinat Protein,Human Frizzled-2 FZD2 (24-190) Protein, Fc Tag,Human Frizzled-2, Fc Tag (FZ2-H5255) is expressed from human 293 c ells (HEK293). It contains AA Gln 24 - Pro 190 (Accession Q14332-1).This protein carries a human IgG1 Fc tag at the C 1 * 1 mg
Catalog # ACRBFZ2-H5255-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Frizzled-2 FZD2 (24-190) Protein, Fc Tag,Human Frizzled-2, Fc Tag (FZ2-H5255) is expressed from human 293 c ells (HEK293). It contains AA Gln 24 - Pro 190 (Accession Q14332-1).This protein carries a human IgG1 Fc tag at the C 1 * 1 mg
Catalog # ACRBFZ2-H5255-1MG
Supplier:  ACROBIOSYSTEMS