ContactUsLinkComponent
Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accessi on P00533-1). 1 * 1 mg
Catalog # ACRBEGR-H522A-1MG
Supplier:  ACRO Biosystems
Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accessi on P00533-1). 1 * 1 mg
Catalog # ACRBEGR-H522A-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together