ContactUsLinkComponent
Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession P24394 -1). 1 * 100 µG
Catalog # ACRBILR-H5253100UG
Supplier:  ACROBIOSYSTEMS
Human IL-4 R alpha, Fc Tag (ILR-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession P24394 -1). 1 * 100 µG
Catalog # ACRBILR-H5253100UG
Supplier:  ACROBIOSYSTEMS