ContactUsLinkComponent
Recombinat Protein,Human SOD1 Cu-Zn SOD Protein, His Tag,Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession AAH01034).This protein carries a polyhistidine tag at the N-terminus. The protein has 1 * 100 µG
Catalog # ACRBSO1-H5148100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human SOD1 Cu-Zn SOD Protein, His Tag,Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession AAH01034).This protein carries a polyhistidine tag at the N-terminus. The protein has 1 * 100 µG
Catalog # ACRBSO1-H5148100UG
Supplier:  ACRO Biosystems
Frequently Bought Together