Biotinylated Human Mesothelin, Fc,Avitag (MSN-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (A ccession AAH09272.1). 1 * 200 µG
Catalog # ACRBMSN-H82F6200UG
Lieferant: ACRO Biosystems
Frequently Bought Together