ContactUsLinkComponent
Human B7-H7, Fc Tag (B77-H5257) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession Q9UM44-1). 1 * 100 µG
Catalog # ACRBB77-H5257100UG
Supplier:  ACRO Biosystems
Human B7-H7, Fc Tag (B77-H5257) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession Q9UM44-1). 1 * 100 µG
Catalog # ACRBB77-H5257100UG
Supplier:  ACRO Biosystems
Frequently Bought Together