ContactUsLinkComponent
Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a calc 1 * 50 µG
Catalog # ACRBUC1-H5140-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human UCH-L1 Protein, His Tag,Human UCH-L1, His Tag (UC1-H5140) is expressed from E.coli cells. It contains AA Me t 1 - Ala 223 (Accession NP004172).This protein carries a polyhistidine tag at the N-terminus. The protein has a calc 1 * 50 µG
Catalog # ACRBUC1-H5140-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together