ContactUsLinkComponent
Biotinylated Human BTLA (31-150), Fc,Avitag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1). 1 * 25 µG
Catalog # ACRBBTA-H82F3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human BTLA (31-150), Fc,Avitag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1). 1 * 25 µG
Catalog # ACRBBTA-H82F3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together