ContactUsLinkComponent
Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession NP000 408). 1 * 1 mg
Catalog # ACRBILA-H52H9-1MG
Supplier:  ACRO Biosystems
Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession NP000 408). 1 * 1 mg
Catalog # ACRBILA-H52H9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together