ContactUsLinkComponent
Cynomolgus PVRIG, Fc Tag (PVG-C5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession A0A2K5WV V8-1). 1 * 1 mg
Catalog # ACRBPVG-C5259-1MG
Supplier:  ACRO Biosystems
Cynomolgus PVRIG, Fc Tag (PVG-C5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession A0A2K5WV V8-1). 1 * 1 mg
Catalog # ACRBPVG-C5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together