ContactUsLinkComponent
Recombinat Protein,Mouse PVRIG Protein, Fc Tag,Mouse PVRIG, Fc Tag (PVG-M5257) is expressed from human 293 cells (HEK293). It contai ns AA Ser 35 - Asp 165 (Accession A0A1B0GS01-1).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBPVG-M5257100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse PVRIG Protein, Fc Tag,Mouse PVRIG, Fc Tag (PVG-M5257) is expressed from human 293 cells (HEK293). It contai ns AA Ser 35 - Asp 165 (Accession A0A1B0GS01-1).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBPVG-M5257100UG
Supplier:  ACROBIOSYSTEMS