ContactUsLinkComponent
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK2 93). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 1 mg
Catalog # ACRBTNA-H526A-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK2 93). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 1 mg
Catalog # ACRBTNA-H526A-1MG
Supplier:  ACROBIOSYSTEMS