ContactUsLinkComponent
Rhesus macaque CD4, Fc Tag (CD4-C5256) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8 ). 1 * 1 mg
Catalog # ACRBCD4-C5256-1MG
Supplier:  ACRO Biosystems
Rhesus macaque CD4, Fc Tag (CD4-C5256) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8 ). 1 * 1 mg
Catalog # ACRBCD4-C5256-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together