Auftragserfassung
Germany
ContactUsLinkComponent
Recombinat Protein,Mouse IgG1 Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession AAK53870.1).This protein carries no tag. The protein has a calculated MW o 1 * 200 µG
Catalog # ACRBIG1-M5208200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse IgG1 Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession AAK53870.1).This protein carries no tag. The protein has a calculated MW o 1 * 200 µG
Catalog # ACRBIG1-M5208200UG
Supplier:  ACROBIOSYSTEMS