ContactUsLinkComponent
Recombinat Protein,Human LILRB1 CD85j ILT2 Protein, Fc Tag,Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK29 3). It contains AA Gly 24 - His 458 (Accession NP006660).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBCDJ-H5252100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human LILRB1 CD85j ILT2 Protein, Fc Tag,Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK29 3). It contains AA Gly 24 - His 458 (Accession NP006660).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBCDJ-H5252100UG
Supplier:  ACROBIOSYSTEMS