ContactUsLinkComponent
Human Fas Ligand, His Tag (active trimer) (MALS verified) (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession AAH17502.1). 1 * 1 mg
Catalog # ACRBFAL-H5241-1MG
Supplier:  ACRO Biosystems
Human Fas Ligand, His Tag (active trimer) (MALS verified) (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession AAH17502.1). 1 * 1 mg
Catalog # ACRBFAL-H5241-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together