ContactUsLinkComponent
Biotinylated Human PD-L1, His,Avitag (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Access ion Q9NZQ7-1). 1 * 200 µG
Catalog # ACRBPDL-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human PD-L1, His,Avitag (PDL-H82E4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Access ion Q9NZQ7-1). 1 * 200 µG
Catalog # ACRBPDL-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together