ContactUsLinkComponent
Recombinat Protein,Human CD14 Protein, Fc Tag,Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession P08571-1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 100 µG
Catalog # ACRBCD4-H5252100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD14 Protein, Fc Tag,Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession P08571-1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 100 µG
Catalog # ACRBCD4-H5252100UG
Supplier:  ACROBIOSYSTEMS