ContactUsLinkComponent
Recombinat Protein,FITC-Labeled Human EGFRvIII Protein, His Tag,FITC-Labeled Human EGFRvIII Protein, His Tag (EGI-HF2H7) is expresse d from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession NP001333870.1). It is the FITC labeled fo 1 * 200 µG
Catalog # ACRBEGI-HF2H7200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,FITC-Labeled Human EGFRvIII Protein, His Tag,FITC-Labeled Human EGFRvIII Protein, His Tag (EGI-HF2H7) is expresse d from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession NP001333870.1). It is the FITC labeled fo 1 * 200 µG
Catalog # ACRBEGI-HF2H7200UG
Supplier:  ACROBIOSYSTEMS