ContactUsLinkComponent
Centrifree Ultrafiltration Device with Ultracel PL membrane 1 * 50 ST The Centrifree® Ultrafiltration device with Ultracel regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samp 1 * 1 ST
  MILX4104
 :  Merck
Centrifree Ultrafiltration Device with Ultracel PL membrane 1 * 50 ST The Centrifree® Ultrafiltration device with Ultracel regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samp 1 * 1 ST
  MILX4104
 :  Merck
Frequently Bought Together