ContactUsLinkComponent
DR.MAISCH REPROSIL-PUR 100 BASIC-C18, 2.5 ΜM, 150 X 3 MM HPLC-SÄULE 1 * 1 items
Catalog # MAISR125.B9.S1503
Supplier:  DR MAISCH
DR.MAISCH REPROSIL-PUR 100 BASIC-C18, 2.5 ΜM, 150 X 3 MM HPLC-SÄULE 1 * 1 items
Catalog # MAISR125.B9.S1503
Supplier:  DR MAISCH
Frequently Bought Together