cNL-B48-Gli1-rv, HPLC, special, quantity delivered min 10 nmol, dry, QC: Mass Check, RNA Oligo Sequence: 5- cag agc ccc cca gca ucg cga gca uca gcg uuc gau gcu ucc gac uaa uca gcc aua uca gcu uac gac ua -3, (No. Bases: 71) 1 * 1 ST
Catalog # MSPPCNLB48GLI1RV
Supplier: MARKET SOURCE PART PROCESS