Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR®, Small sample viscosity standards

VWR®, Small sample viscosity standards

2,884 cSt, Glass bottle, ASTM D2162, 60 mL
Catalog # 99360.600
Supplier:   VWR Chemicals
VWR®, Small sample viscosity standards
Catalog # 99360.600
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Small sample viscosity standards
VWR®, Small sample viscosity standards

2,884 cSt, Glass bottle, ASTM D2162, 60 mL

CAS Number:  
Price
48,50 €
Log in for availability
1 * 60 mL
Product Details & Documents

These standards are certified in strict accordance with ASTM D2162 at 40 and 100 °C under ISO 17025 and ISO 17034 accreditation, and have been manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids by Automated Houillon Viscometer and other similar type test equipment.

  • Fully traceable to international standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Certified in strict accordance with ASTM D2162
  • 60 ml pack is a lower volume, cost effective solution
  • Tamper-evident security packaging

Certifications: ISO 17025 and ISO 17034 accreditation.

Caution: The viscosity value given in the ordering table below is the nominal kinematic viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    60 mL
  • Viscosity at 40 °C:
    2,884 cSt
  • Test method:
    ASTM D2162

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.