27 components 10mg/l each of Ag Al B Ba Be Bi Cd Ce Co Cr Cs Cu Fe La Li Mn Ni P Pb Rb Sr Te Th Tl U V Zn in HNO3 1% . Duree de conservation : 6 mois a ouverture du flacon, Duree de validite: 12 mo 1 * 500 mL
Catalog # CMPLRM037631L5
Supplier: CUSTOM MADE CHEMICALS LAB