Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FUROSINE DEDICATED COLUMN 250X4.6MM 5µM 1 * 1 items
Catalog # MAIS5133663
Supplier:
DR MAISCH
Expand image
FUROSINE DEDICATED COLUMN 250X4.6MM 5µM 1 * 1 items
Catalog # MAIS5133663
Supplier:
DR MAISCH