Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tauwasserabflussheizung230 Volt, 6,5 WattDurchmesser 8mmLänge 1700mm 1 * 1 ST
Catalog # GRAM765071698
Supplier:
GRAM
Expand image
Tauwasserabflussheizung230 Volt, 6,5 WattDurchmesser 8mmLänge 1700mm 1 * 1 ST
Catalog # GRAM765071698
Supplier:
GRAM