PoQ_biotin_Oligo2, HPLC, special, quantity delivered min 2.000 nmol, dry, QC: Mass Check, DNA Oligo Sequence: 5'- Biotin-CCA CTG ACT GTA CGA TGA AGA TGC TGA CGA GCT C -3', (No. Bases: 34) 1 * 2 Nmo
Catalog # MSPPPOQ_BIOTIN-34
Supplier: MARKET SOURCE PART PROCESS