Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN NUCLEOSIL 100 5 C8 250 X 4.0 MM 1 * 1 items
Catalog # HISMMZ3025-250040
Supplier:
HICHROM
Expand image
COLUMN NUCLEOSIL 100 5 C8 250 X 4.0 MM 1 * 1 items
Catalog # HISMMZ3025-250040
Supplier:
HICHROM