Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant CD40L Cytokine, source: E. coli 1 * 10 µG
Catalog # INVVRCYEC-HCD40L
Supplier:
Invivogen
Expand image
Recombinant CD40L Cytokine, source: E. coli 1 * 10 µG
Catalog # INVVRCYEC-HCD40L
Supplier:
Invivogen