Conductivity Standard 12880 uS/cm at 25C CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 500 mL
Conductivity Standard 12880 uS/cm at 25C CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 500 mL