RP246_AN_Atto550_3, HPLC, special, quantity delivered min 10 nmol, dry, QC: Mass Check, DNA Oligo 247,60 EUR Sequence: 5'- GCT CTG A TG CCG CAT AGT TAA GCC AGC CCC GAC ACC CG -Atto 550 -3', (No. Bases: 38) 1 * 1 ST
Catalog # MSPPRP246ANATTO
Supplier: MARKET SOURCE PART PROCESS