ContactUsLinkComponent
Setaflesh Series 3e ActiveCool Operation Temperature range 10 to 135 C Units of temperature C F Ignitor Electric Cup Closed Sample cup material Aluminium Heating/cooling method Peltier cell Measurement time 1 or 2 minutes Sample volume 2 ml or 4 1 * 1 ST
Catalog # STAN33220-0
Supplier:  STANHOPE SETA
Setaflesh Series 3e ActiveCool Operation Temperature range 10 to 135 C Units of temperature C F Ignitor Electric Cup Closed Sample cup material Aluminium Heating/cooling method Peltier cell Measurement time 1 or 2 minutes Sample volume 2 ml or 4 1 * 1 ST
Catalog # STAN33220-0
Supplier:  STANHOPE SETA
Frequently Bought Together