ContactUsLinkComponent
100 KEMIO SENSORS FOR CHLORINE TESTING, STANDARD RANGE. FREE CHLORINE RANGE: 0.2 – 10 MG/L. TOTAL CHLORINE RANGE: 0.2 – 75 MG/L 1 * 100 items
Catalog # PALIKEM21CLO
Supplier:  Palintest
100 KEMIO SENSORS FOR CHLORINE TESTING, STANDARD RANGE. FREE CHLORINE RANGE: 0.2 – 10 MG/L. TOTAL CHLORINE RANGE: 0.2 – 75 MG/L 1 * 100 items
Catalog # PALIKEM21CLO
Supplier:  Palintest
Related Information