Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MAGNIFIER LED 20DPT. 5X DIAM. 58 MM 1 * 1 items
Catalog # ESCH15115
Supplier:
ESCHENBACH OPTIK
Expand image
MAGNIFIER LED 20DPT. 5X DIAM. 58 MM 1 * 1 items
Catalog # ESCH15115
Supplier:
ESCHENBACH OPTIK