ContactUsLinkComponent
Desiccator novus , borosilicate glass. with safety protefan coating as implosion and splinter protection with needle valve stopcock in side tubulature and knobbed lid. 1 * 1 items
Catalog # WITG5.070.200
Supplier:  witeg Labortechnik
Desiccator novus , borosilicate glass. with safety protefan coating as implosion and splinter protection with needle valve stopcock in side tubulature and knobbed lid. 1 * 1 items
Catalog # WITG5.070.200
Supplier:  witeg Labortechnik