Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PHARMAFLUIDICS µPACTM COLUMN, 200CM LENGTH, WITH PILLAR ARRAY BACKBONE AT INTERPILLAR DISTANCE OF 2.5 µM, C18
Catalog # MSPP5525031518200
Supplier:
MARKET SOURCE PART PROCESS
Expand image
PHARMAFLUIDICS µPACTM COLUMN, 200CM LENGTH, WITH PILLAR ARRAY BACKBONE AT INTERPILLAR DISTANCE OF 2.5 µM, C18
Catalog # MSPP5525031518200
Supplier:
MARKET SOURCE PART PROCESS