Objectives for Rebel Microscopes
Plan apochromat, N.A. 0,08, 6,2 mm
Catalog # ECHLRMAG9022
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Magnification:2×
- Description:Plan apochromat, N.A. 0,08
- Seq:22
- Working distance:6,2 mm