Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat oil, N.A. 1,25, 0,2 mm
Catalog # ECHLRMAG1002
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG1002
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan achromat oil, N.A. 1,25, 0,2 mm
Price
284,00 €
Log in for availability
1 * 1 ST
Specifications
  • Magnification:
    100×
  • Description:
    Plan achromat oil, N.A. 1,25
  • Seq:
    05
  • Working distance:
    0,2 mm